DNAPac™ PA200 Nucleic Acid Columns, Thermo Scientific
Proveedor: Thermo Scientific
088781 063421 062998 063419 063423 088780 063000 063425
10044-492EA 29407.5 MXN
10044-488 10044-490 10044-350 10044-494 10044-492 10044-476 10044-352 10044-474
DNAPac™ PA200 Nucleic Acid Columns, Thermo Scientific
Columnas de cromatografía
For ultra-high-resolution HPLC analysis of synthetic oligonucleotides, use Thermo Scientific™ DNAPac PA200 columns.
These small particle size, strong anion-exchange columns resolve full length oligonucleotides from n–1, n+1, and other failure sequences not possible with other columns. Control the retention times and selectivity by choice of salt, pH, and solvent, and tailor the separation to the requirements of your different oligonucleotide analysis challenges.