Reacti-Vial™ Small Reaction Vials
Proveedor: Thermo Scientific
Reacti-Vial™
TS-13221 TS-13222 TS-13099 TS-13220 TS-13100 TS-13225 TS-13223 TS-13097
10055-170EA 9100.34 MXN
10055-170 10055-148 10055-166 10055-168 10055-234 10055-150 10055-172 10055-152
Reacti-Vial™ Small Reaction Vials
Viales
These small reaction vials have an internal cone designed to make small-sample collection and handling easy and convenient. The cone feature is particularly useful for removing small quantities of sample with a syringe.
  • Extra thick glass wall magnifies the sample ideal for observing chemical reactions
  • Amber vials are light-sensitive to well protect the compounds
These vials are ideal for residue isolation, derivative preparation, maximum sample retrieval, moisture protection, sample storage, purification, precipitations, centrifugations and solvent separations.
Información de suministro: Vials are supplied complete with open-top screw caps and teflon/rubber laminated discs.