ContactUsLinkComponent
Pearl® Adjustable Volume Micropipettes, Single Channel
Pearl® Adjustable Volume Micropipettes, Single Channel
Catalog # 10127-192
undefined
Pearl® Adjustable Volume Micropipettes, Single Channel
Catalog # 10127-192
Supplier Number:  HS120104
Specifications
  • Volume:
    100 - 1000 µl
  • Cat. no.:
    10127-192
  • Increments:
    0.02, 0.02, 0.02 µl
  • Color Code:
    Royal blue
  • Supplier No.:
    HS120104
Specifications
About this item
A complete range of single channel pipettor volumes to cover all your possible needs.
  • Internal metal parts for increased accuracy and calibration stability
  • Easy volume adjustment with locking tab to prevent accidental change
  • Large digital volume display
  • All Pipettes come with a separate tip ejector
The sleek erogonomic design and reduced plunger pressure help reduce the chance of RSI, as the pipettors require 2/3rd the operation pressure of most standard pipettors. All devices feature internal metal parts for increased accuracy and calabiration stability, easy volume adjustment with locking tab to prevent accidental change, and separate tip ejector and a large digital volume display.
Certifications: Each pipettor is tested in accordance with Q/TFBM02 2002 and ISO 8655 standards and a calibration certificate is supplied. Manufactured in accordance with ISO9001 and GLP regulations. CE, UKCA
Frequently Bought Together