OTAL Foot Pumps, Bürkle
Proveedor: Bürkle
5000-4016 5000-8012 5000-6000 5000-1800 5000-3012 5000-2200 5000-5018 5000-1500
10216-944EA 4785.63 MXN
10216-944 10216-946 10216-940 10216-942 10216-950 10216-948 76501-446 10216-938
OTAL Foot Pumps, Bürkle
Bombas Bombas para bidón
Delivery tubes made of PP are resistant against weak acids, caustic solutions and detergents.
Delivery tubes made of PVDF are especially for acids and caustic solutions; not for strongly oxidizing acids and caustic solutions.
Delivery tubes made of stainless steel 304 are for solvents and liquids of VbF Hazard Class AIII (not easily flammable).
Battery pump for safe filling of car batteries with acid or distilled water.
  • Simple handling
  • Very good pump capacity due to large volume foot bellows
  • With universal bungs made of soft PVC, for container openings of 30 - 70 mm Ø