Control Seal Labels, Bürkle
Proveedor: Bürkle
5303-0014 5303-9100 5303-0016 5303-1014 5303-0011 5303-9000 5303-1011 5303-1016
10216-978PK 2748.25 MXN
10216-978 10216-996 10216-986 10217-340 10216-992 10216-982 10217-000 10217-342
Control Seal Labels, Bürkle
Etiquetas
Designed to seal the holes in sacks or boxes created when samples are taken, the strong adhesive on the ‘close-it®’ label allows adhesion to surfaces that are slightly moist, frozen or coated with powder. The aluminium backing creates a vapor barrier and seals the container hermetically.
  • Adheres extremely firmly
  • Can be written on
  • Color coded for easy recognition
Envase: Rolls contain 500 (95×95 mm) or 250 (150×150 mm) stickers.