Benchtop Draining Racks, Bürkle
Proveedor: Bürkle
9640-3050 9640-3040 9640-3060
10217-322EA 11545.29 MXN
10217-324 10217-322 10217-320
Benchtop Draining Racks, Bürkle
Gradillas Soportes para drenaje
  • Stable bench top rack, can use both sides
  • Rods are simply clicked into place
  • Collecting pan can be easily removed for emptying
The stable rods are individually placed into the holes of the draining rack and are clicked into place. Their positions can be changed at any time. Using a combination of rods of three different lengths (60 mm, 100 mm and extension to 150 mm), the draining rack can be adapted to any apparatus which is to be dried.
These draining racks are made of PVC.