VWR® Tissue Culture Plates, Non-Treated, Sterilized, Non-Pyrogenic
Proveedor: Avantor
TCP001006 TCP001001 TCP001012 TCP001096 TCP002096 TCP001024 TCP001004 TCP001384 TCP001048
10861-562CS 3553.84 MXN
10861-556 10813-630 10861-558 10861-562 10861-564 10861-554 75780-348 10861-560 10814-224
VWR® Tissue Culture Plates, Non-Treated, Sterilized, Non-Pyrogenic
Microplacas Placas de cultivo celular
Cell and tissue culture plates are ideal for cell growth and cell yields.
  • Available with 8 different growth surface areas
  • Uniform well volume
  • Flat well bottom and round bottom plates available
  • Well surface is smooth and free from striation
  • Alphanumerically coded for easy identification
  • Suitable for all common instruments and automation
  • Sterilized by electron beam irradiation
  • Non-pyrogenic
Raised rims on wells with the uniform rings on the lid help to reduce evaporation.
Plates are designed for use in applications where cell attachment is not desired. They are available in individual recyclable PET, with an operating temperature range of −20 to +50 °C (−4 to +122 °F). Plates are not autoclavable. Single-position lid reduces the risk of cross-contamination and handling mistakes.
Increased cell attachment surface: This highly hydrophilic surface offers a significant advantage over the traditional cell culture surface. The surface treatment can improve cell spreading and attachment, and is suitable for cells that may adhere poorly due to cell phenotype, stressful culture conditions, or those which normally require additional biological coatings for attachment.
TC-treated: These dishes are vacuum-gas plasma or corona treated for consistent cell attachment and growth.
Non-treated: Non-treated cell and tissue culture dishes are ideal for applications where cell attachment is not desired.
Certificaciones: ISO 13485 2003 and ISO 9001 2008 certified.
Envase: Individually packaged in peel-to-open paper/plastic blister pack. Every case is printed with a Lot Number for quality traceablity.