VWR® Sample Sak™ Sample Bags
Proveedor: Avantor
VWR® Sample Sak™
740-101 740-100
11216-002CS 7658.57 MXN
11216-002 11216-000
VWR® Sample Sak™ Sample Bags
Bolsas Bolsas para muestras
Ideal collection/transport containers for powder and granular samples including grain, seeds, feeds, coffee, powdered soaps, and even damp soil.
  • For solid and semi-solid samples
  • Imprinted writing area
  • Tin tie tabs
Heat-sealed bags are constructed of 50 lb. machine finished white Kraft paper with tin tie tabs. Lined with 1.25 mm polyethylene. Imprinted on each bag is a personalized area for recording sample data and date.