VWR® Microcentrifuge Tube Rack, PP
Proveedor: VWR
VWR®
C8210-VWR
20901-675PK 674.38 MXN
20901-675
VWR® Microcentrifuge Tube Rack, PP
Gradillas Tube Racks
Each polypropylene microcentrifuge tube rack stores 20 standard 1.5–2.0 mL microcentrifuge tubes in clearly numbered cavities.
  • Numbered cavities for quick identification
  • One-handed transportation
  • Autoclavable
  • Polypropylene construction
Easily transportable with one hand. Made of autoclavable polypropylene. Non-skid rubber feet.