KIMAX® Chromatographic Columns, Kimble Chase
Proveedor: DWK Life Sciences
KIMAX®
17800-22300 17800-19300 17800-19500 17800-11500 17800-22500 17800-11300
21503-123CS 2088.63 MXN
21503-123 21503-156 21503-067 21503-065 21503-010 21503-015
KIMAX® Chromatographic Columns, Kimble Chase
Columnas de cromatografía
With removable stopcock of PTFE and replaceable glass tip.
Lower end of the column is tooled for extra strength and tight, leakproof fit. Glass tip is supplied.