Corning® 96-Well Plates, Polypropylene, Without Lids
Proveedor: Corning
Corning®
3363 3343 3344 3364 3357 3359 3356 3355 3365
29444-102CS 9300 MXN
29444-102 80081-120 29444-100 29445-112 29444-400 29444-104 29445-150 80081-118 29445-152
Corning® 96-Well Plates, Polypropylene, Without Lids
Microplacas Microplacas HTS
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or non sterile
Información para pedidos: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µl plate is packaged 10 plates per bag and 5 bags per case.