Corning® 96-well Clear Polystyrene Microplates, Corning
Proveedor: Corning
Corning®
3369 3697 3798 3590 3997 3599 3366 3585 3591 3360 3300 3841 3797 7007 3595 3358 3788 3367 3361 9018 3370 3368 3695 9018BC 3797 3628 3690 3641 3474 3596 3896 3799 3795 3898 3897 3696 3598 3628BC 3894 9017
29442-316CS 2700 MXN
29442-316 25381-048 29444-288 29442-314 29444-320 29442-398 29442-058 71001-358 66025-626 29442-318 470211-270 29445-154 29442-070 89501-318 29442-396 29442-056 29442-054 29442-392 29442-050 29442-404 29444-454 29442-402 29442-400 29442-068 29444-456 29442-322 29444-316 29445-142 89089-826 29443-954 29443-034 25381-050 89092-706 29442-066 25381-052 25381-054 29442-062 25381-056 29444-274 29442-060
Corning® 96-well Clear Polystyrene Microplates, Corning
Microplacas Placas de cultivo celular
Cell culture microplates are sterile and nonpyrogenic. Lids available where indicated.
NA Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays. NA Cat. Nos. 3585 and 3595 have special low evaporation lids. NA Cat. No. 3841 is aseptically manufactured.