VWR® Lead Ring Flask Weights
Proveedor: VWR
VWR®
80018-0118 80018-0119 80018-0117 80018-0116
29700-060EA 1035.05 MXN
29700-060 29700-048 29700-026 29700-004
VWR® Lead Ring Flask Weights
Pesas Pesas de matraces
Protect and stabilize expensive glassware with Vikem® vinyl-coated lead rings.
  • Stabilize glassware immersed in water baths
  • Reduce floating, breakage and chemical spills
  • Coated with bright red Vikem® vinyl
  • Chemical-resistant
  • Will not scratch flasks
  • Available in five diameter/weight combinations
Vikem® vinyl coated lead rings stabilize glassware immersed in water baths, thereby reducing floating, breakage, and chemical spills.