J.T.Baker® Plate SRW Magnetic
Proveedor: Ritter, part of Avantor
J.T.Baker®
43001-05061 43001-05051 43001-05041
43001-0506CS 5398.66 MXN
43001-0506 43001-0504 43001-0505
J.T.Baker® Plate SRW Magnetic
Microplacas Microplacas HTS
These plates are manufactured from high purity PP for efficient sample preparation and are suitable for use with magnetic particle processors such as the KingFisher™ Flex 96 and Presto.
  • Standard SBS footprint
  • Cylindrical wells enable good sample mixing
  • Slightly raised rims reduce the risk of cross-contamination
  • Alphanumeric code assists registration and identification of individual samples