Loading Dye
Proveedor: ALDON CORP SE
LL0248-5ML
470024-814EA 69.12 MXN
470024-814
Loading Dye
Tintes Colorantes para electroforesis
Solution used for tracking DNA during agarose electrophoresis; prepared with ultra-pure water and reagent-grade chemicals. 10X concentration.