1000 Watt Type 3 Voltage Converter, Step Up and Step Down Voltage Transformer
Proveedor: INTERNATIONAL VIDEO & ELECTRONICS
DS-1000
470061-810EA 3817.98 MXN
470061-810
1000 Watt Type 3 Voltage Converter, Step Up and Step Down Voltage Transformer
Fuentes de alimentación
The International DS-1000 Diamond Series 1000 Watt Type 3 Voltage converters offer best in class performance in voltage transformers. They are thoroughly tested to be free of defects in workmanship and materials and hand soldering insures to meet U.S Military specifications.
  • USA grounded male plug
  • Extra adapter included to use with 220 V outlets
  • Converts 110/120 V up to 220/240 V and vice versa
  • Works anywhere in the world
  • Heavy duty for continuous use
  • Extra durable lifetime coil is included
  • Recommended for high value and/or sensitive items
Weight: 15 lbs. (6.8 kg)
Dimensions: 7.25x8.25x5.75" (18.4x21x14.6 cm)
Ideal when using a ¼" drill, a clothes dryer or an LCD TV or when moving or visiting other countries. BEST Diamond series 5 year warranty.