VWR® Pipette Fillers
Proveedor: Avantor
HS20632A
470123-222CS 231.41 MXN
470123-222
VWR® Pipette Fillers
Pipetas Bombas de pipeteadores
Pipette fillers eliminate the hazardous practice of drawing liquids into pipettes by mouth.
  • Filters work with pipettes up to 50 ml in size
  • Filters help avoid sample contamination
  • Safe, precise delivery
  • Ideal for volatile, corrosive, or poisonous liquids
  • Fits a standard serological pipet
To use, insert the pipette into the bottom opening of the filler, draw to the desired volume, and expel by pressing lettered valves.
The filters are made of soft rubber. Colors may vary. Dimensions: 54×54×130 mm (L×W×D).