Order Entry
PowerSpin™ DXB Horizontal Spin Centrifuge
PowerSpin™ DXB Horizontal Spin Centrifuge
Certificates
undefined
PowerSpin™ DXB Horizontal Spin Centrifuge
Specifications
  • Catalog No:
  • 470013-944
  • 470013-946
  • 470144-636
  • Max. speed:
  • 500~5500 rpm
  • Max. RCF:
  • 4000 ×g
  • Tube capacity:
  • 2-15 mL
Specifications
Product Information
  • Catalog #
    Supplier #
    Supplier No.
    Availability
    Price
  • PowerSpin™ DXB Horizontal Spin Centrifuge
    PowerSpin™ DXB Horizontal Spin Centrifuge
    470013-946
    C8706
    C8606
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $76,938.54
    $76,938.54
    Each (1UN)
       $76,938.54
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Max. speed
    500~5500 rpm
    Max. RCF
    4000 ×g
    Tube capacity
    2-15 mL
  • 6 Place Swing Out Rotor for 2-15 mL Tubes for Horizontal Spin Centrifuge
    6 Place Swing Out Rotor for 2-15 mL Tubes for Horizontal Spin Centrifuge
    470013-944
    C8600-01
    C8600-01
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $23,267.90
    $23,267.90
    Each (1UN)
       $23,267.90
    Specifications:
    Specifications
    Environmentally Preferable
    Max. speed
    Max. RCF
    Tube capacity
  • Tube Shield for Horizontal Spin Centrifuge
    Tube Shield for Horizontal Spin Centrifuge
    470144-636
    C800-02
    C800-02
    Log in for availability
    Log in for availability
    Paquete de 2
    Paquete de 2 - $591.11
    $591.11
    Paquete de 2
       $591.11
    Specifications:
    Specifications
    Environmentally Preferable
    Max. speed
    Max. RCF
    Tube capacity