Burner Wing Top
Proveedor: VWR International
H-6650 C-1505/1 H-6650
470148-920CS 1624.31 MXN
470202-860 470148-920 18026-108
Burner Wing Top
Calentadores Mecheros de gas
It provides a broad, flat flame and fits mixing tubes with 1/2" outer diameter without stabilizer or 11/16" with stabilizer.