Needle Valve-Controlled Burner
Proveedor: VWR International
C1475/1
470149-470CS 6025.82 MXN
470149-470
Needle Valve-Controlled Burner
Calentadores Mecheros de gas
Efficient and Durable for General Lab Use.
  • Base Diameter: 4"
  • Height: 6"
  • Mixing Tube Diameter: ¹/₂"
  • Natural gas
  • 7.6 mm inner diameter tube
Easily cleaned metal orifice for natural gas with a heat value of 1000 to 1050 BTU. Gas flow is controlled by a needle valve located above the base, permitting easy flame adjustment and minimizing possibility of tipping. Air flow is adjusted by a heavy gauge mixing tube threaded into a heavy die-cast zinc alloy base. With hose barb for connection to 6 mm tubing.