Order Entry
GreatNeck® Slip Joint Pliers
GreatNeck® Slip Joint Pliers
  470149-880
undefined
GreatNeck® Slip Joint Pliers
  470149-880
 :  9002E
$122.58
  Each (1UN)
  • Description:
    Pliers with 5" Slip Joint
  • Cat. no.:
    470149-880
  • Weight:
    0.30 lbs.
  • Supplier No.:
    9002E
These pliers are designed for gripping small objects and tightening fasteners. Constructed with drop forged steel for strength, and feature a milled jaw for added gripping power.
  • Ergonomic comfort grip handles for reduced user fatigue
  • 36 Piece Display Set