/
Mexico
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Ratchet Headgear Systems, 3M™
55666-304
:
3M OCCUPATIONAL HEALTH
Ratchet Headgear Systems, 3M™
55666-304
:
3M OCCUPATIONAL HEALTH
:
7000002290
$761.04
Select uom
Unit of Measure
Each (1UN)
-
+
Color:
Blue
Description:
Ratchet Headgear H8A
Cat. no.:
55666-304
Supplier No.:
82501-00000