Order Entry
SP Bel-Art Vikem® Pipette Filler,Vinyl, Bel-Art Products, a part of SP
  57497-306
undefined
SP Bel-Art Vikem® Pipette Filler,Vinyl, Bel-Art Products, a part of SP
  57497-306
 :  F37886-0000
$1,406.21
Select uom
Unit of Measure
Paquete de 6
  • Ø:
    5 cm
  • Description:
    Pipette Filler
  • Length:
    10 cm
  • Cat. no.:
    57497-306
  • Supplier No.:
    378860000
This pipette aid is operated by squeezing then releasing the bulb to aspirate fluid into the pipet.
  • Vikem® vinyl with a tapered polyethylene chuck
  • Designed for quick removal when filling is complete
  • Simple to use
After aspirating the fluid, the pipette aid is then removed, and the pipette top quickly covered with a gloved finger to control and dispense fluid.