VWR® Cuvette Racks, PP
Proveedor: VWR
VWR®
F18516-0787 F18516-0000
58020-104CS 4494.52 MXN
58020-104 58020-105
VWR® Cuvette Racks, PP
Gradillas Soportes de cubetas
These two-tier racks support cuvettes with a minimum of physical contact, greatly reducing the chance of damage.
  • Racks accommodate 12 cuvettes of 10 mm light path
  • Racks will not scratch quartz cuvettes
  • Rack 58020-105 features Stack Rack™ Interlocking end plates for secure stacking
  • Manufactured from polypropylene, and steam Autoclavable at 121°C (250°F)
Only the corners of the cuvettes touch the racks, minimizing chance for contamination or damage.