VWR® Spinbar® Stir Bars, Octagon, Color-Coded
Proveedor: VWR
VWR® Spinbar®
37109-6016 37109-6001 37109-6623 37109-6024 37109-6033 37109-6007 37109-6031 37109-6004 37109-6013 37109-6009 37109-6008 37109-6032 37109-6010 37109-6012 37109-6015 37109-6011 37109-6022 37163-6001 37109-6018 37109-6014 37109-6017
58949-046CS 2541 MXN
47751-474 58949-046 47751-472 58949-058 58949-048 70670-098 58948-089 58948-087 58948-085 58949-053 58949-064 58949-042 58949-054 58949-044 58949-066 58949-056 58949-060 58949-050 58949-051 58949-062 58949-052
VWR® Spinbar® Stir Bars, Octagon, Color-Coded
Imánes de agitación Barras magnéticas de agitación
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Pivot ring aids in reducing friction and chattering
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
Available in red, yellow or blue. All stir bars have a pivot ring with the exception of the 12.7 x 3mm sizes (58949-044, -046, and -050). Autoclavable.
Choose from red, yellow or blue.
Información para pedidos: Assortment includes six of size 1 x 5/16" (25 x 7mm), 2 each color; Yellow, Red, Blue.