Nunc-Immuno™ MicroWell™ 96-Well Plates, Thermo Scientific
Proveedor: Thermo Scientific
Nunc-Immuno™ MicroWell™
446612 436110 430341 467340 446140 467340 449824 436110 475094 456537 456537 437796 437112 460984 456529 430341 475094 436111 439454 439454 442404 475434 437112 449824 436111 467320 446612 437111 475434 460984 437111 467320 456529 442404 446140 437796
62409-314CS 10720.02 MXN
CA41321-033 CA62409-005 CA62409-004 62409-314 CA62409-003 CA62409-442 CA62409-002 CA62409-024 CA62409-046 CA62409-440 CA62407-160 62409-052 62407-164 43300-454 73520-830 CA43300-440 43300-456 CA62409-314 62409-005 62409-004 73520-832 62409-003 62409-046 62409-002 62409-024 62407-162 CA62409-052 62407-160 CA62409-050 62409-050 CA62407-164 CA62407-162 43300-440 43300-442 CA43300-456 CA43300-454
Nunc-Immuno™ MicroWell™ 96-Well Plates, Thermo Scientific
Microplacas Microplacas para inmunología
One-piece 96-well MicroWell™ plates are designed for colorimetric, fluorescent, and luminescent immunoassays and binding assays
Assay plates are ideal for colorimetric ELISA tests and are frosted for easy handling and labeling. LumiNunc plates are designed for luminescent assays, but can also be used for fluorescent assays. FluoroNunc plates are for fluorescent assays.
Certified plates guarantee uniform binding capacity of CV less than 5%, intra well O.D. within ±10% from the mean of the plate. Background reading for all wells within ±0.005 adsorbance units from mean. Made of virgin polystyrene.
Plates are made from a highly controlled and characterized resin. Four surfaces are offered to optimize mobilization. The MaxiSorp™ surface is a hydrophilic/hydrophobic mix that binds to a wide variety of biomolecules, but has not been optimized to bind IgG. The PolySorp™ surface is hydrophobic and is ideal for the immoblization of hydrophobic biomolecules such as lipids and hydrophobic proteins. MediSorp™ plates are less hydrophilic than MaxiSorp™, but more hydrophilic than PolySorp™ surfaces. This surface binds many biomolecules that have a mixed hydrophilic/hydrophobic character, including immunoglobulins. Non-specific binding is reduced for samples that contain serum or plasma. The MultiSorp™ surface is more hydrophilic than MaxiSorp™ plates. This surface binds more hydrophilic proteins such as glycoproteins. It also has high affinity for polar lipids.
Información para pedidos: Packed five plates per sleeve. The 400 µL non-certified MaxiSorp™ Plate is bar coded. For lids and sealing films, see 62409-118 series.