Nunc-Immuno™ and Fluoronunc™/Luminunc™ 96-Well Modules and Frames, Thermo Scientific
Proveedor: Thermo Scientific
Nunc-Immuno™ Fluoronunc/Luminunc™
468667 473709 475078 467466 437591 467466 467140 469264 437591 473709 469957 469949 467120 437702 469957 446471 469078 475086 444865 475523 469922 445101 475523 473717 473717 441653 469078 475515 437702 446473 445101 469914 467120 444865 469922 437915 469264 475086 437915 469914 475078 441653 475515 467140 468667 469949 463200
62409-261CS 9770.11 MXN
CA62409-306 CA62409-228 CA62409-448 62409-316 CA62409-302 CA62409-300 CA62409-289 62409-137 CA62409-288 62409-136 CA62409-287 62409-135 62409-134 62409-133 62409-132 CA62409-261 CA62409-283 62409-130 62409-295 62409-294 CA62409-280 62409-280 89094-310 CA62409-316 62409-306 62409-228 CA62409-137 CA62409-136 62409-302 CA62409-135 CA62409-134 CA62409-133 62409-289 62409-300 89094-304 62409-288 CA62409-132 62409-287 CA62409-450 89094-306 CA62409-130 CA62409-295 CA62409-294 62409-283 62409-261 73520-764 73520-762
Nunc-Immuno™ and Fluoronunc™/Luminunc™ 96-Well Modules and Frames, Thermo Scientific
Microplacas Microplacas para inmunología
Composed of individual modules consisting of either 8, 12, or 16 wells, these plates are designed specifically for quantitative immunoassays and solid phase assays
Well modules and frames are compatible with 96-well plate automation. Well modules are held firmly by the frame and will not come loose when inverted. Modules are made of virgin polystyrene. The mixed hydrophilic/hydrophobic MaxiSorp™ surface binds to a wide variety of biomolecules, but has been optimized to bind IgG. Recommended for assays where glycoproteins, such as antibodies, are adsorbed into the plastic. The hydrophobic PolySorp™ surface is recommended for the immobilization of hydrophobic biomolecules, such as lipids and hydrophobic proteins. PolySorp™ is less polar and can be used for viral membrane antigens. MaxiSorp™ and PolySorp™ plates are certified for consistent binding of IgG. The MediSorp™ surface is recommended for adsorption of medium to large biomolecules with hydrophilic and hydrophobic properties, such as immunoglobulins, peptides, and albumins. Non-specific binding is reduced for samples that contain serum or plasma. Well modules and frames are designed for one-way fit; plates comply with SBS standards for 96-well microplates. Choose from round-, C-, flat-, or StarWell™-bottom wells. The C-bottom wells combine the readability of a flat-bottom well with the washing ease of a round-bottom well. The StarWell™-bottom design increases the surface area to volume ratio, which decreases assay times and increases sensitivity.
The MultiSorp™ surface is recommended for adsorption of glycoproteins, polar lipids, and phospholipids.
Nunc-Immuno™ modules feature high optical quality, an alphanumeric orientation system, and are ideal for colorimetric ELISA tests.
FluoroNunc™/LumiNunc™ modules are ideal for fluorescent and luminescent assays. LockWell™ modules use a locking well frame to prevent jamming.
Certified plates guarantee uniform binding capacity and a CV <5% between wells, with all results being within ±10% from the mean of the plate and background reading for all wells within ±0.005 adsorbance units from mean.
Información para pedidos: Wells are available in strips of 8, 12, or 16 as complete plates with frames. Clear strips of 8 or 16 are available separately.