Aluminum Seals, Tear-Out, Kimble Chase
Proveedor: DWK Life Sciences
73820-20 73820B-13 73820B-20 73820C-20 73820D-13 73820C-13 73820D-20 73820-13
66010-797CS 3343.64 MXN
66004-030 66004-032 66010-795 66010-797 66004-028 66004-034 66004-024 66004-026
Aluminum Seals, Tear-Out, Kimble Chase
Tapas Tapones para viales de cromatografía
These tear-out seals are designed to secure septa and stoppers to bottles and vials.
  • Tamper-evident center tear-out feature protects target area
  • For use with 13 and 20 mm serum finished bottles and vials
  • Allow for easy access to septa