WHEATON® Vial Racks, DWK Life Sciences
Proveedor: DWK Life Sciences
868806 985750 868804 868805 868808 868810 985800
66022-478CS 5257.84 MXN
66022-580 66022-525 66022-569 66022-478 66022-523 66022-556 14007-178
WHEATON® Vial Racks, DWK Life Sciences
Gradillas Gradillas de viales criogénicos
Manufactured from polypropylene, vial racks are easy to clean and autoclavable
Each rack features alphanumeric indexing to simplify identification and selection. The racks are sturdy and stackable, even when vials are in place.
Información para pedidos: When selecting the proper rack, the vial O.D. cannot exceed the well I.D. dimension.