Aluminum Crimp Seals with Prefitted Septa, Thermo Scientific
Proveedor: Thermo Scientific
C4011-7G C4011-1AP C4011-98R C4011-98B C4011-3A C4011-5A C4011-98G C4011-6A C4011-4A C4011-7A C4011-2A C4011-7R C4011-98Y C4011-7B C4011-1A
66064-192PK 588.3 MXN
66065-166 66064-192 66030-144 66030-140 66030-148 66030-150 66030-142 66065-418 66011-186 66064-224 66011-184 66065-162 66030-146 66065-164 66011-182
Aluminum Crimp Seals with Prefitted Septa, Thermo Scientific
Tapas Tapones para viales de cromatografía
Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on
Used for primarily for routine analysis. Offer moderate resealability and excellent chemical inertness.