CHEMWARE® PTFE Watch Glasses, Saint Gobain Performance Plastics
Proveedor: Saint-Gobain
D1069016 D1069013 D1069014 D1069012 D1069015
66111-024EA 797.87 MXN
66111-024 66111-046 66111-002 66111-068 66111-080
CHEMWARE® PTFE Watch Glasses, Saint Gobain Performance Plastics
Vidrios de relojes
Ultra-shallow, concave dish is ideal for holding, weighing, crystallizing, evaporating and drying small samples of liquids, powders and solids.
  • Shatter-proof PTFE material
  • Inert chemically resistant
  • Smooth and unbreakable
  • Easy to clean
It can also be used as a convex cover for beakers and erlenmeyer flasks to limit airborne contaminants, while still allowing gas exchange. All sizes are molded to the same contour for easy stacking.