RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
72830-022
:
- Pack type:Squeeze bottle
- Description:RNase AWAY®
- Size:250 ml
- Cat. no.:72830-022
- Supplier No.:7000
RNase AWAY® eliminates RNase and DNA from laboratory surfaces. The surface decontaminant is ideal for decontaminating apparatus, benchtops, glassware, and plastic ware. The decontaminant leaves no residue to interfere with gel polymerization or staining and contains no strong acids.
Note: Should not be used as a reagent.
Note: Should not be used as a reagent.
The surface decontaminant is ideal for decontaminating apparatus, benchtops, glassware, and plastic ware. The decontaminant leaves no residue to interfere with gel polymerization or staining and contains no strong acids