Nunc® 96-Well Polystyrene Conical Bottom Microwell Plates, Thermo Scientific
Proveedor: Thermo Scientific
249940 249952 249940 249935 249935 249952
73520-134CS 25467.62 MXN
CA73520-134 89027-038 CA89027-036 73520-134 CA89027-038 89027-036
Nunc® 96-Well Polystyrene Conical Bottom Microwell Plates, Thermo Scientific
Microplacas Placas de cultivo celular
Nunc® 96-well polystyrene conical plates are optimized for robotics, automated readers and liquid handling systems
The plates are clear which makes them ideal for optical measurements. The lids in the package come with rings which prevent cross contamination. The well shape of these plates maximizes sample recovery.
Certificaciones: All microplates manufactured by Nunc® meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)