Drying/Heating Ovens with Natural Convection, Avantgarde.line ED Series, BINDER
Proveedor: Binder
9010-0336 9010-0340 9010-0334
75783-622EA 2625.8 MXN
75783-622 75783-630 75783-626
Drying/Heating Ovens with Natural Convection, Avantgarde.line ED Series, BINDER
Hornos Hornos de convección natural y con ventilación
BINDER Avantgarde.Line ED Drying ovens are ideal for sterilization, drying, and thermal storage tasks which do not require high drying rates or special time parameters.
  • Ovens provide reliable, reproducible, quick task results
  • Easy to clean
BINDER ovens provide fast uniform drying for drying and sterilization applications. The Advanced Preheating Technology (APT.line™) provides temperature accuracy of 1°C with a wide temperature range from 5°C to 15°C above ambient to 300 °C (depending on the model size). With the natural convection and high rate of air exchange, thermal process run with increased efficiency.
Certificaciones: UL.
Información para pedidos: The ovens include two chrome-plated shelves. For additional sizes not listed below, please refer to the BINDER Classic ED Drying Ovens.