VWR® Standard Line Sterile Centrifuge Tubes with Flat or Plug Caps, PP
Proveedor: VWR
VWR®
CTNA301 CTNA314 CTNA214 CTNA305 CTNA312 CTNA211 CTNA311 CTNA209 CFT115500 CTNA213 CTNA216 CTNA207 CTNA210 CTNA303 CFT114500 CTNA208 CTNA313 CFT113500 CTNA215 CTNA304
10026-078CS 2337.21 MXN
10026-078 76204-402 76204-404 10026-074 75845-202 10026-076 75845-206 10025-702 75845-204 10025-712 10025-700 10025-688 10025-698 10025-686 10025-696 10025-694 76204-542 10025-692 76204-400 10025-690
VWR® Standard Line Sterile Centrifuge Tubes with Flat or Plug Caps, PP
Tubos Tubos de centrífuga
These disposable conical-bottom and freestanding tubes are manufactured from high-quality USP Class 6 gamma-resistant polypropylene.
  • Available in two volumes: 15 and 50 ml
  • Available with two bottom styles: conical bottom and self-standing bottom
  • Longer length screw caps with sealing ring prevent leakage
  • Easy-to-read black graduations are accurate to ±2%, with 1 ml increments (15 ml) or 2.5 ml increments (50 ml)
  • Large, unerasable frosted white writing area
  • Both the graduations and writing areas are chloroform-resistant
  • Engraved graduation at the conical bottom of each tube and leak-proof
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
They feature black printed graduations and smooth inner walls for easy filling and sample preparation. Tubes are RNase-free, DNase-free, and non-pyrogenic.
Caps are available in two styles: flat, with a smooth surface for additional labeling options, and plug style for a secure seal when using shakers and horizontal incubators. All caps are made of white high-density polyethylene.
A white printed writing area on the side allows for permanent coding. Tubes and caps are autoclavable at 121 °C and freezable at –80 °C.
Max. rotation speed up to 12500×g for conical bottom tubes, and 10000×g for self-standing tubes.
Certificaciones: ISO 13485:2003 and ISO 9001:2008.
Información para pedidos: All tubes are packaged with caps attached and sterilized with irradiation. Plastic racks are available as optional accessories in seven colors. An assorted color pack offers one of each color: blue, green, orange, red, and yellow. Racks are designed for use with plug or flat bottom tubes. Cap and tube packed separately for items 75845-202, 75845-204, and 75845-206.
Envase: Racked tubes are packaged in environmentally responsible (fully recyclable) chipboard racks or plastic racks. Bagged tubes are packaged in durable and re-sealable (zip closure) bags. Inner packaging is individually labelled for lot-to-lot traceability.