DR300 Pocket Colorimeters, Hach
Proveedor: Hach
LPV445.97.65110 LPV445.97.10110 LPV445.97.09110 LPV445.97.12110 LPV445.97.60110 LPV445.97.15110 LPV445.97.25110 LPV445.97.50110 LPV445.97.16110 LPV445.97.06110 LPV445.97.52110 LPV445.97.00110 LPV445.97.02110 LPV445.97.40110 LPV445.97.51110 LPV445.97.01110 LPV445.97.62110 LPV445.97.26110 LPV445.97.22110 LPV445.97.04110 LPV445.97.00110 LPV445.97.03110
76311-198EA 27630.12 MXN
76311-076 76311-110 76311-198 76311-074 76311-184 76311-196 76311-072 76311-084 76311-194 76311-070 76311-082 76311-192 76311-206 76311-204 76311-202 76311-078 76311-068 76311-200 76311-080 76311-190 76318-832 CA76318-832
DR300 Pocket Colorimeters, Hach
Colorímetros Colorímetros monoparamétricos
Proven past. Innovative future.
  • Simple, intuitive operation reduces potential manual error, ensuring accurate measurement data you can trust, time after time
  • Larger display with improved backlight makes reading measurements in all conditions even easier
  • Single parameter go-anywhere portable photometer
  • Battery operation for a maximum of 5000 tests
  • Larger, better display
  • Data connectivity-Bluetooth to Claros (bluetooth connectivity currently available only in US, Canada and EU)
The tried and true Hach pocket colorimeter II has been used by hundreds of thousands of water professionals in harsh conditions for almost two decades. Key mechanical improvements: upgraded waterproofing; larger display; ergonomic design. Infused with the smart capabilities that modern technology has to offer: optional bluetooth connectivity with automatic data transfer, and seamless system integration. The Hach DR300 Pocket Colorimeter: the next generation in portable instrumentation. Reduce data collection hassles, eliminate transcription errors, and ensure stronger compliance traceability.
Connected - Optional bluetooth connectivity allows you to safely transfer measurement data to your existing database or to Claros-Hach®'s water intelligence system - reducing errors and saving time by eliminating manual data entry.
Reliable - Rugged, waterproof design withstands whatever conditions you encounter in the field (drops, extreme temperatures, rain and dirt) and still delivers years of dependable, accurate measurements.
Certificaciones: Waterproof instrument IP 67 (even better than PCII).
Información para pedidos: Includes DR300 pocket colorimeter, manual, sample cells, and a carrying case. Reagents much be ordered separately. DR300 Pocket Colorimeters are not sold with chemical reagents.