Blowers for Laboratory Hoods, Labconco®
Proveedor: Labconco
71810003726 7182200 706920010814 706840010814 71816003726 17725110010814 706940010814 71814003726 706860010814 71800003726 71808003726 17725120010814 7180400 706880010814 17725010010814 71812003726 7180600 17705110010814 706800010814 706960010814 7182000 706820010814 71802003726 706900010814
82006-778EA 96808.76 MXN
77296-336 77296-334 82006-790 82006-792 77296-332 82006-780 77296-330 82006-754 82006-786 82006-742 82006-756 82006-778 82006-744 82006-788 82006-794 82006-750 97019-806 82006-782 82006-796 82006-752 82006-784 82006-740 82006-746 82006-748
Blowers for Laboratory Hoods, Labconco®
Ventiladores
Fiberglass blowers are recommended for fume hood exhaust systems in moderate to highly corrosive conditions
Coated steel blowers are recommended for low to moderately corrosive applications and have a phenolic coating on impeller and housing.
Both blowers have a dry powder epoxy-coated base and weather cover for protection in extreme weather. Sheaves are adjustable to allow for air volume adjustment. Self-adjusting gravity belt tightener ensures consistent performance because belt is held taut by weight of motor. Motor mount keeps the motor in place should the belt ever fail.
The housing is made from molded fiberglass-reinforced polyester for durability and long life. Impellers are made of injection-molded polypropylene to resist corrosive atmospheres.
Información para pedidos: Items indicated below as REDISHIP units can be shipped within 48 hours of receiving your order.