VWR® Floating Foam Tube Racks
Proveedor: Avantor
VWR®
HS2165AV HS2166V HS2165BV HS2165CV
82017-634CS 117521.25 MXN
82017-634 82017-632 82017-630 82017-628
VWR® Floating Foam Tube Racks
Gradillas Tube Racks
These high-density polyethylene foam racks are ideal for floating tubes in water baths, tubs, or beakers.
  • Resistant to water absorption
  • Color-coded for easy identification
  • Different shapes available
Closed cell foam is resistant to water absorption so the racks can be washed and reused. Foam handles make it easy to lift samples out of baths. Racks come in different colors and shapes for easy identification. Round rack 82017-634 is sized to fit a beaker and features unique x-shaped slits that accommodate a variety of tube sizes.