VWR® 96-Place Microtube Racks
Proveedor: Avantor
VWR®
HS2345CV HS2345FV HS2345BV HS2345AV HS2345EV HS2345DV
82024-494CS 4919.38 MXN
82024-490 82024-494 82024-484 82024-492 82024-488 82024-486
VWR® 96-Place Microtube Racks
Gradillas Tube Racks
These polypropylene racks can accommodate ninety-six 0.5 mL microcentrifuge tubes on one side or 1.5 to 2.0 mL microcentrifuge tubes on the opposite side in a standard 8x12 format.
  • Polypropylene construction
  • Wells are more visible and tubes are easier to place
Racks feature easy-to-grasp tabs on each end and a handy snap-in-place lid. Grid lines are molded on the side of the rack that accommodates the 0.5 mL tubes and raised rings are molded on the other side to make the wells more visible and tubes easier to replace.
Información para pedidos: Assorted rack pack 82024-494 includes one each of blue, green, pink, yellow, and orange.