VWR® Large-Volume, Heavy-Walled Bulbs
Proveedor: VWR
VWR®
HS20630AV HS20630CV HS20630DV HS20630BV HS20630EV
82024-568CS 19983.6 MXN
82024-568 82024-566 82024-572 82024-570 82024-564
VWR® Large-Volume, Heavy-Walled Bulbs
Pipetas Bombas de pipeteadores
Easy-to-grip bulbs feature heavy-walled necks for use with pipettes and syringes.
  • Heavy-walled design
  • Natural rubber fabrication
  • Easy to grip
Made of natural red rubber. The 150 ml bulb (82024-572) features reinforcing ribs to aid in larger volume applications.