VWR® Safety Bulb Pipette Filler
Proveedor: VWR
VWR®
HS20623V
82024-574CS 55123.29 MXN
82024-574
VWR® Safety Bulb Pipette Filler
Pipetas Bombas de pipeteadores
Designed for use with small and large pipettes, bulb features a white plastic attachment with a silicone conical adapter for safe and secure attachment to pipettes.
  • Easy application
  • Secure attachment to pipettes
  • Simple operation
  • Suction action for drawing liquids
Attachment does not require twisting or rough pipette handling. Simply squeeze bulb and gently press pipette to form seal. Liquid is drawn by suction; lift away bulb to control fluid dispensing with index finger. Supplied with elastic band for hanging on bottles or wrist.