VWR® Sterile Sample Bags with Specimen Sponge
Proveedor: Avantor
VWR®
KSS-61105-VW1 KSS-61100-VW1 KSS-61100-VW1
89003-806CS 17040.96 MXN
89003-806 89085-576 CA89003-806
VWR® Sterile Sample Bags with Specimen Sponge
Bolsas Bolsas para muestras
Sterile sample bags are ideal for surface sampling in a wide range of applications.
  • Sterile
  • Includes cellulose sponge
  • Polyethylene sterile tubing
  • Write-on strip
  • Wire closure for an airtight seal
  • Wide-sealed bottoms for leakproof protection
They provide a reliable method for monitoring potentially hazardous environments for the presence of microorganisms as well as foodborne pathogens.
Each bag includes a cellulose sponge that measures 3.8Wx7.6Lx1.6Hcm (11/2x3x5/8") when hydrated. Choose from a biocide-free, dry sponge or a foil-sealed sponge premoistened with a sterile buffer solution.
Bags are made of virgin polyethylene tubing with no side seals. They feature a wire closure for an airtight seal, wide-sealed bottoms for added leakproof protection, and a write-on strip. They are easy to use: tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal.
Precaución: Do not use with alcoholic substances or where temperature exceeds 100°C (212°F).