VWR® Water Sampling Kits
Proveedor: VWR
VWR®
KWS-22200-VW1 KWS-21100-VW1 KWS-22200-VW1 KWS-21100-VW1
89003-816CS 1702.47 MXN
89003-816 CA89003-818 89003-818 CA89003-816
VWR® Water Sampling Kits
Bolsas Bolsas para muestras
These sterile sampling bags are ideal for collecting chlorinated water specimens.
  • Sterile polyethylene construction
  • Suited for water sampling
  • Easy to use
  • Wire closure provides an airtight seal
They are made of sterile virgin polyethylene tubing with no side seals.
Bags feature a wire closure for an airtight seal and wide-sealed bottoms for added leakproof protection. They are easy to use: simply tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal.
UPA-approved for drinking water analysis.
Bags include a nontoxic, nutritive tablet containing 10 mg of active sodium thiosulfate.