VWR® Poly Bags, Pre-Opened
Proveedor: VWR
VWR®
2715
89071-416CS 3137.12 MXN
89071-416
VWR® Poly Bags, Pre-Opened
Bolsas Bolsas de almacenamiento
Pre-opened bags on rolls are for use on automated packing machines.
  • Suitable for use with automated packing machines
  • Tubular construction
  • Durable bottom seal
Bags have a slit in front and a perforated back for automatic loading or fast hand loading of products. Made with a tubular construction and a durable bottom seal. Wound on a 8.9 cm (3¹/₂") core. Core plug has a 2.5 cm (1") diameter hole.