VWR® Pink Anti-Static Bags
Proveedor: VWR
VWR®
12210 12030
89072-822CS 4392.88 MXN
89072-766 89072-822
VWR® Pink Anti-Static Bags
Bolsas Bolsas para muestras
This premium anti-static bag won't corrode polycarbonate.
  • Antistatic properties
  • Amine-free
  • Performs in low humidity
With a surface resistivity less than 1×1012 ohms/sq VWR's 4 mm bags. This can also be heat sealed, tied, stapled or taped shut.
Certificaciones: Meeta the full static decay requirements of MIL-PRF-81705E, Type II and EIA-541.