Gas Dispersion Tubes, Ace Glass Incorporated
Proveedor: Ace Glass
7202-04 7202-42 7202-20 7202-36 7202-40 7202-34 7202-32 7202-14 7202-16 7202-46 7202-10 7202-12 7202-18 7202-48 7202-02 7202-08 7202-06 7202-50 7202-44 7202-38
89083-492EA 1336.57 MXN
89083-488 89083-498 89083-500 89083-502 89083-480 89058-370 89083-490 89083-482 89058-372 89083-492 89058-360 89058-362 89083-486 89058-354 89058-364 89083-496 89058-356 89058-366 89058-358 89058-368
Gas Dispersion Tubes, Ace Glass Incorporated
Tubos per gas
Gas dispersion tubes feature a porous fritted glass tip
The O.D. for all sizes is 10mm, and all tubes fit a No. 11 Ace-Thred. Manufactured from borosilicate glass.