VWR® Sterile Sample Bags
Proveedor: VWR
VWR®
EFR-1015-VW1 EPR-7012-VW1 EPL-3070-VW1 EPL-5515-VW1 BFR-5590-VW1 EFL-4590-VW1 EPR-7012-VW1 EPL-4590-VW1 EPR-4590-VW1 EPR47012-VW1 EFL4712S-VW1 EDL41216-VW1 BFR-4590-VW1 EFR-5590-VW1 EFL47012-VW1 EPL-7012-VW1 EPR-3070-VW1 EDL41216-VW1 EPL45590-VW1 EFL-1015-VW1 EFL47012S-VW1 BPL-5515-VW1 SCL-2424-VW1 EPL-4512-VW1 SCL33030-VW1 BFL-5515-VW1 EFR-1012-VW1 EPL-5590-VW1 EPL-4590-VW1 EFR-1015-VW1 BFL-5590-VW1 EPR47012-VW1 EDL41212-VW1 EDL41218-VW1 BPL-7012 EPR-4590-VW1 EFR-1012-VW1 EFL-5590-VW1 EPL47012-VW1 BPR-5590-VW1 BPR-7012-VW1 EFR-4590-VW1 SCL33037-VW1 EPR-5590-VW1 EFR47012-VW1 EFL-1012-VW1 BPR-7012-VW1 EPL-5590-VW1 BFL45590-VW1 SCL21520-VW1 EDL41212-VW1 BPR-4590-VW1 SCL21520-VW1 EPL-4575-VW1 BPL-5590-VW1 EPL-5515-VW1 EPR-3050-VW1 BPL-5590-VW1 BPR-5590-VW1 BPL-4512-VW1 BFL45590-VW1 EFR-3070-VW1 EPR-5590-VW1 EDL41224 EPR-7012S-VW1 EPL-7012-VW1 EFL-1012-VW1 EPR-4512-VW1 EPR-3070-VW1 EDL41218-VW1 EDL41224-VW1 BFL-4512-VW1 EFR-5590-VW1 BPL-4590-VW1 EFL-1015-VW1 BFR-5590-VW1 EFR47012-VW1 EFL-5515-VW1 EFL-4590-VW1 BPR-4590-VW1 EPL45590-VW1 EFR-4590-VW1 EDL41524-VW1 EPL47012-VW1 EPR45590-VW1 EPR-3050-VW1 EPL-7015-VW1 EFL-5590-VW1 EPL-3070-VW1
89004-418CS 5269.02 MXN
89085-556 89004-418 89085-558 CA83007-556 82007-700 89085-552 CA89085-560 89085-554 76289-492 CA89085-528 82007-708 CA83007-558 82007-706 89000-644 89000-648 89004-420 89085-550 89004-422 CA89000-650 89000-640 89004-424 89000-642 82007-696 83007-556 CA89004-418 89000-190 CA89085-532 83007-558 CA83007-560 CA89085-530 CA83007-562 82007-698 CA82007-706 CA89085-538 89085-528 CA89000-642 CA82007-708 89000-636 CA89000-644 89000-638 CA89000-648 83007-560 CA89004-420 89085-560 CA89004-422 CA89000-640 CA89004-424 CA82007-694 CA89085-542 76196-376 89085-535 83007-562 89085-530 CA82007-698 CA89085-540 CA89000-190 89085-532 76196-372 82007-726 82007-724 89085-538 89085-537 82007-728 CA89085-546 76265-604 CA89000-636 CA89000-638 CA89085-580 76196-370 CA89085-556 89085-544 89085-546 89085-540 76214-710 CA89085-552 CA89085-550 89085-542 CA82007-724 82007-714 CA82007-722 82007-718 89085-548 CA82007-726 82007-716 CA89085-558 10818-386 89085-580 89000-650 76221-334
VWR® Sterile Sample Bags
Bolsas Bolsas para muestras
These sample bags are ideal for transportation and storage of solids, semisolids, and liquids for environmental and carcass sampling, biomedical and pharmaceutical research, quality assurance procedures, food industry applications, and clinical and veterinary medicine.
  • Pliant polyethylene sterile tubing
  • Highly resistant
  • Wire closure for an airtight seal
  • Round wire bags have two wires for closure
Bags are made of pliant, highly resistant virgin polyethylene sterile tubing with no side seals. They feature a wire closure for an airtight seal, and wide-sealed bottoms for added leakproof protection. To use, tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal.
Note: Do not use with alcoholic substances or where temperature exceeds 100 °C (212 °F).
Certificaciones: Comply with EPA, FDA, USDA, and HACCP regulatory and industry requirements.
Información para pedidos: Bags 89085-560 and 76289-492 are packaged in a double pouch for cleanroom applications.