VWR® Refrigerated Circulating Baths
Proveedor: VWR
AP20R-30-V11B TMPVP07D4SC TMPVP15P3 AD15R-30-V11B TMPVP15D3 MX07R-20-V11B TMPVP07M2LC TMPVP07P2SC TMPVP28D3 TMPVP07P2S AP15R-30-V11B TMPVP45D2C TMPVP45P2C TMPVP28D3C AD20R-30-V11B TMPVP20D3 TMPVP15P3C TMPVP07M2S TMPVP20D3C TMPVP07M2SC TMPVP07P4S AD15R-40-V11B AD28R-30-V11B TMPVP20P3 TMPVP07D2LC TMPVP15D3C AP07R-40-V11B TMPVP45P2 TMPVP07D2L TMPVP15P4C TMPVP07D4S AP15R-40-V11B TMPVP28P3C AP28R-30-V11B AD07R-40-V11B AP07R-20-V11B AP45R-20-V13D TMPVP07P4SC TMPVP07M2L TMPVP20P3C TMPVP07D2SC AD7LR-20-V11B AD07R-20-V11B TMPVP15D4C TMPVP28P3 TMPVP07D2S TMPVP45D2 TMPVP15P4 MX7LR-20-V11B TMPVP15D4
89203-006EA 251773.99 MXN
89202-990 10017-324 10017-288 10017-322 89202-994 10017-286 10017-320 89202-998 10017-318 10017-316 10017-314 89203-006 89203-002 10017-312 10017-310 89202-962 89202-966 10017-308 89202-958 10017-306 10017-304 89203-014 10017-664 10017-302 89203-010 89202-970 10017-662 10017-300 10017-660 89202-974 10017-658 10017-336 10017-656 10017-294 10017-292 10017-290 89202-982 89203-022 10017-334 10017-654 10017-332 10017-652 10017-330 89202-986 89202-978 10017-648 10017-646 10017-328 10017-644 10017-326
VWR® Refrigerated Circulating Baths
Termostatos con circulación Sistemas de circulación cerrada Circuladores de enfriamiento
VWR® Refrigerated Circulating Baths offer precise temperature control, at temperature ranges from -40° to 200°C (depending on the controller).
  • Offer precise temperature control
  • Available in a wide range of sizes
  • Three controller options
Available in a wide range of sizes, with three controller options, these refrigerated circulating baths are ideal for many applications.
The large, LCD display on the MX temperature controller is easily viewed across the lab. The Advanced Digital temperature controller offers a touch-pad, while the Advanced Programmable Temperature Controller includes a full touch-screen display, and programmability (ten 100-step programs).
The Advanced Series Controllers offer Swivel 180™ technology, which allows you to rotate the controller independently from the bath. All heated circulating baths include LidDock™ lid stowing system and DuraTop™, the precision molded phenolic bath top, which remains cooler at high temperatures and resists laboratory chemicals.
Certificaciones: ETL listed. CE marked. MX Controller complies with DIN 12876-1 Class I safety requirements for use with non-flammable liquids. Advanced Digital and Advanced Programmable comply with DIN 12876-1 Class III safety requirements for use with flammable liquids. cETLus compliant.
Información para pedidos: Refrigerated Circulators include reservoir cover; bypass tubing; male inlet and outlet adapters for 3/8, 1/4" and 3/16" (.95, .63 and .47 cm) tubing. 1/4" NPT to M16 adapters also included on 50 Hz models.