Viscosity Standard Oils for Rotational Viscometers, Fungilab
Proveedor: Fungilab
V12Z22 V12Z50 V12Z23 V12Z34 V12Z46 V12Z26 V12Z30 V12Z42
89237-082EA 4036.5 MXN
89237-074 89237-082 89237-072 89237-080 89237-092 89237-090 89237-088 89237-078
Viscosity Standard Oils for Rotational Viscometers, Fungilab
Viscosity standard oil for checking the viscosity measurements for rotational viscometers.
Specifically formulated for use with equipment with minimal temperature control. Supplied in 600 ml 'ready to use' packs specially designed to eliminate cleaning of laboratory glassware, which is inherently difficult due to the nature of silicone. These range of standards provide both calibration and verification options for rotational viscometer test equipment.
Certificaciones: Calibration Certificate included at two temperatures: 20 °C and 25 °C (68 °F and 77 °F). ISO 17025 / ISO Guide 34 dual certified general-purpose viscosity standards are tested in accordance with ASTM D 2162.