VWR® Aluminum Test Tube Racks
Proveedor: VWR
11648-300 11648-301 11648-299
89259-896EA 754.17 MXN
89259-896 89259-898 89259-894
VWR® Aluminum Test Tube Racks
Gradillas Tube Racks
VWR® Aluminum Test Tube Racks are punched from hard sheet aluminum.
  • Edges and holes have a smooth finish
  • Practical, rust-proof design
S-shaped rack is practical and rust-proof. The low profile and wide, flat base prevent rack from tipping. Rack also slides into other racks for compact storage.